Assay Design



Gene Transcripts

Gene Symbol
VBA4
Entrez Gene ID
851697
Gene Name
Gene Aliases
YDR119W
Chromosome Location
Chr.IV: 688227 - 690533 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180427.1 NP_010404.1 1 - 1 2152 95

View Full Product Details