Assay Design
Gene Transcripts
- Gene Symbol
- MKC7
- Entrez Gene ID
-
851722
- Gene Name
- Gene Aliases
-
YDR144C, YPS2
- Chromosome Location
-
Chr.IV: 744311 - 746101 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180451.3 |
NP_010428.3 |
1 - 1 |
1654 |
82 |
View Full Product Details