Assay Design
Gene Transcripts
- Gene Symbol
- GIR2
- Entrez Gene ID
-
851730
- Gene Name
- Gene Aliases
-
YDR152W
- Chromosome Location
-
Chr.IV: 765706 - 766503 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180459.3 |
NP_010436.3 |
1 - 1 |
654 |
85 |
View Full Product Details