Assay Design
Gene Transcripts
- Gene Symbol
- AHA1
- Entrez Gene ID
-
851800
- Gene Name
- Gene Aliases
-
YDR214W
- Chromosome Location
-
Chr.IV: 892875 - 893927 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180522.3 |
NP_010500.3 |
1 - 1 |
950 |
77 |
View Full Product Details