Assay Design
Gene Transcripts
- Gene Symbol
- ADR1
- Entrez Gene ID
-
851802
- Gene Name
- Gene Aliases
-
YDR216W
- Chromosome Location
-
Chr.IV: 895035 - 899006 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180524.3 |
NP_010502.3 |
1 - 1 |
3747 |
101 |
View Full Product Details