Assay Design
Gene Transcripts
- Gene Symbol
- COX20
- Entrez Gene ID
-
851817
- Gene Name
- Gene Aliases
-
YDR231C
- Chromosome Location
-
Chr.IV: 926293 - 926910 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180539.1 |
NP_010517.1 |
1 - 1 |
543 |
70 |
View Full Product Details