Assay Design
Gene Transcripts
- Gene Symbol
- DON1
- Entrez Gene ID
-
851866
- Gene Name
- Gene Aliases
-
YDR273W
- Chromosome Location
-
Chr.IV: 1010176 - 1011273 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180581.3 |
NP_010559.3 |
1 - 1 |
950 |
67 |
View Full Product Details