Assay Design
Gene Transcripts
- Gene Symbol
- SWR1
- Entrez Gene ID
-
851934
- Gene Name
- Gene Aliases
-
YDR334W
- Chromosome Location
-
Chr.IV: 1135932 - 1140476 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180642.1 |
NP_010621.1 |
1 - 1 |
4450 |
80 |
View Full Product Details