Assay Design
Gene Transcripts
- Gene Symbol
- ATP17
- Entrez Gene ID
-
851983
- Gene Name
- Gene Aliases
-
YDR377W
- Chromosome Location
-
Chr.IV: 1228611 - 1228916 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180685.3 |
NP_010665.3 |
1 - 1 |
246 |
86 |
View Full Product Details