Assay Design
Gene Transcripts
- Gene Symbol
- ARO10
- Entrez Gene ID
-
851987
- Gene Name
- Gene Aliases
-
YDR380W
- Chromosome Location
-
Chr.IV: 1234218 - 1236125 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180688.3 |
NP_010668.3 |
1 - 1 |
1853 |
89 |
View Full Product Details