Assay Design
Gene Transcripts
- Gene Symbol
- SXM1
- Entrez Gene ID
-
852004
- Gene Name
- Gene Aliases
-
KAP108, YDR395W
- Chromosome Location
-
Chr.IV: 1263324 - 1266158 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180703.3 |
NP_010683.3 |
1 - 1 |
2550 |
79 |
View Full Product Details