Assay Design
Gene Transcripts
- Gene Symbol
- PPM1
- Entrez Gene ID
-
852045
- Gene Name
- Gene Aliases
-
YDR435C
- Chromosome Location
-
Chr.IV: 1332983 - 1333969 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180743.1 |
NP_010723.1 |
1 - 1 |
747 |
63 |
View Full Product Details