Assay Design
Gene Transcripts
- Gene Symbol
- YHP1
- Entrez Gene ID
-
852062
- Gene Name
- Gene Aliases
-
YDR451C
- Chromosome Location
-
Chr.IV: 1361120 - 1362181 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180759.3 |
NP_010739.3 |
1 - 1 |
949 |
104 |
View Full Product Details