Assay Design
Gene Transcripts
- Gene Symbol
- SNM1
- Entrez Gene ID
-
852089
- Gene Name
- Gene Aliases
-
YDR478W
- Chromosome Location
-
Chr.IV: 1414575 - 1415171 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180786.3 |
NP_010766.3 |
1 - 1 |
448 |
79 |
View Full Product Details