Assay Design
Gene Transcripts
- Gene Symbol
- EMI2
- Entrez Gene ID
-
852128
- Gene Name
- Gene Aliases
-
YDR516C
- Chromosome Location
-
Chr.IV: 1474974 - 1476476 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180824.3 |
NP_010804.3 |
1 - 1 |
1454 |
68 |
View Full Product Details