Assay Design
Gene Transcripts
- Gene Symbol
- IRC22
- Entrez Gene ID
-
856717
- Gene Name
- Gene Aliases
-
YEL001C
- Chromosome Location
-
Chr.V: 150301 - 150978 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178816.1 |
NP_010915.1 |
1 - 1 |
446 |
102 |
View Full Product Details