Assay Design
Gene Transcripts
- Gene Symbol
- GLC3
- Entrez Gene ID
-
856705
- Gene Name
- Gene Aliases
-
GHA1, YEL011W
- Chromosome Location
-
Chr.V: 133120 - 135234 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178826.1 |
NP_010905.1 |
1 - 1 |
2054 |
87 |
View Full Product Details