Assay Design
Gene Transcripts
- Gene Symbol
- EDC3
- Entrez Gene ID
-
856700
- Gene Name
- Gene Aliases
-
DCP3, LSM16, YEL015W
- Chromosome Location
-
Chr.V: 126629 - 128284 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178830.1 |
NP_010901.1 |
1 - 1 |
1539 |
98 |
View Full Product Details