Assay Design
Gene Transcripts
- Gene Symbol
- YAT2
- Entrez Gene ID
-
856745
- Gene Name
- Gene Aliases
-
YER024W
- Chromosome Location
-
Chr.V: 202192 - 204963 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178915.1 |
NP_010941.1 |
1 - 1 |
2649 |
94 |
View Full Product Details