Assay Design
Gene Transcripts
- Gene Symbol
- GLN3
- Entrez Gene ID
-
856763
- Gene Name
- Gene Aliases
-
YER040W
- Chromosome Location
-
Chr.V: 229795 - 231987 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178931.3 |
NP_010958.3 |
1 - 1 |
2051 |
99 |
View Full Product Details