Assay Design
Gene Transcripts
- Gene Symbol
- ACA1
- Entrez Gene ID
-
856769
- Gene Name
- Gene Aliases
-
YER045C
- Chromosome Location
-
Chr.V: 240032 - 241501 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178936.3 |
NP_010964.3 |
1 - 1 |
1353 |
101 |
View Full Product Details