Assay Design
Gene Transcripts
- Gene Symbol
- HMF1
- Entrez Gene ID
-
856785
- Gene Name
- Gene Aliases
-
HIG1, YER057C
- Chromosome Location
-
Chr.V: 270737 - 271126 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178948.3 |
NP_010978.3 |
1 - 1 |
248 |
86 |
View Full Product Details