Assay Design
Gene Transcripts
- Gene Symbol
- HXT10
- Entrez Gene ID
-
850536
- Gene Name
- Gene Aliases
-
YFL011W
- Chromosome Location
-
Chr.VI: 112345 - 113985 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179955.1 |
NP_116644.1 |
1 - 1 |
1547 |
92 |
View Full Product Details