Assay Design
Gene Transcripts
- Gene Symbol
- HIS2
- Entrez Gene ID
-
850581
- Gene Name
- Gene Aliases
-
YFR025C
- Chromosome Location
-
Chr.VI: 203743 - 204750 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179990.1 |
NP_116681.1 |
1 - 1 |
948 |
88 |
View Full Product Details