Assay Design
Gene Transcripts
- Gene Symbol
- OCH1
- Entrez Gene ID
-
852845
- Gene Name
- Gene Aliases
-
LDB12, NGD29, YGL038C
- Chromosome Location
-
Chr.VII: 425362 - 426804 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180903.1 |
NP_011477.1 |
1 - 1 |
1151 |
70 |
View Full Product Details