Assay Design
Gene Transcripts
- Gene Symbol
- GUP1
- Entrez Gene ID
-
852796
- Gene Name
- Gene Aliases
-
YGL084C
- Chromosome Location
-
Chr.VII: 350616 - 352298 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180949.1 |
NP_011431.1 |
1 - 1 |
1553 |
98 |
View Full Product Details