Assay Design
Gene Transcripts
- Gene Symbol
- MMS2
- Entrez Gene ID
-
852793
- Gene Name
- Gene Aliases
-
YGL087C
- Chromosome Location
-
Chr.VII: 346406 - 346904 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180952.1 |
NP_011428.1 |
2 - 2 |
349 |
85 |
View Full Product Details