Assay Design
Gene Transcripts
- Gene Symbol
- CEG1
- Entrez Gene ID
-
852747
- Gene Name
- Gene Aliases
-
YGL130W
- Chromosome Location
-
Chr.VII: 266145 - 267524 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180995.3 |
NP_011385.3 |
1 - 1 |
1251 |
69 |
View Full Product Details