Assay Design
Gene Transcripts
- Gene Symbol
- RTG2
- Entrez Gene ID
-
852640
- Gene Name
- Gene Aliases
-
YGL252C
- Chromosome Location
-
Chr.VII: 25718 - 27484 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181118.1 |
NP_011262.1 |
1 - 1 |
1648 |
69 |
View Full Product Details