Assay Design
Gene Transcripts
- Gene Symbol
- CAX4
- Entrez Gene ID
-
852924
- Gene Name
- Gene Aliases
-
CWH8, YGR036C
- Chromosome Location
-
Chr.VII: 558149 - 558868 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181165.3 |
NP_011550.3 |
1 - 1 |
451 |
92 |
View Full Product Details