Assay Design
Gene Transcripts
- Gene Symbol
- CCM1
- Entrez Gene ID
-
853053
- Gene Name
- Gene Aliases
-
DMR1, RRG2, YGR150C
- Chromosome Location
-
Chr.VII: 790459 - 793053 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181279.1 |
NP_011666.1 |
1 - 1 |
2448 |
93 |
View Full Product Details