Assay Design
Gene Transcripts
- Gene Symbol
- CIR1
- Entrez Gene ID
-
853121
- Gene Name
- Gene Aliases
-
YGR207C
- Chromosome Location
-
Chr.VII: 910843 - 911628 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181336.1 |
NP_011723.1 |
1 - 1 |
643 |
103 |
View Full Product Details