Assay Design
Gene Transcripts
- Gene Symbol
- ECM29
- Entrez Gene ID
-
856355
- Gene Name
- Gene Aliases
-
YHL030W
- Chromosome Location
-
Chr.VIII: 40084 - 45690 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179110.1 |
NP_011833.1 |
1 - 1 |
5549 |
74 |
View Full Product Details