Assay Design
Gene Transcripts
- Gene Symbol
- PIH1
- Entrez Gene ID
-
856429
- Gene Name
- Gene Aliases
-
NOP17, YHR034C
- Chromosome Location
-
Chr.VIII: 176965 - 177999 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179164.1 |
NP_011899.1 |
1 - 1 |
943 |
88 |
View Full Product Details