Assay Design
Gene Transcripts
- Gene Symbol
- RPP1
- Entrez Gene ID
-
856459
- Gene Name
- Gene Aliases
-
YHR062C
- Chromosome Location
-
Chr.VIII: 222877 - 223758 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179192.1 |
NP_011929.1 |
1 - 1 |
750 |
83 |
View Full Product Details