Assay Design
Gene Transcripts
- Gene Symbol
- PEX28
- Entrez Gene ID
-
856555
- Gene Name
- Gene Aliases
-
YHR150W
- Chromosome Location
-
Chr.VIII: 397251 - 398990 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179281.1 |
NP_012020.1 |
1 - 1 |
1652 |
88 |
View Full Product Details