Assay Design
Gene Transcripts
- Gene Symbol
- ERG9
- Entrez Gene ID
-
856597
- Gene Name
- Gene Aliases
-
YHR190W
- Chromosome Location
-
Chr.VIII: 484845 - 486179 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179321.1 |
NP_012060.1 |
1 - 1 |
1247 |
92 |
View Full Product Details