Assay Design
Gene Transcripts
- Gene Symbol
- SCH9
- Entrez Gene ID
-
856612
- Gene Name
- Gene Aliases
-
HRM2, KOM1, YHR205W
- Chromosome Location
-
Chr.VIII: 509363 - 511837 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179336.1 |
NP_012075.1 |
1 - 1 |
2351 |
92 |
View Full Product Details