Assay Design
Gene Transcripts
- Gene Symbol
- CFD1
- Entrez Gene ID
-
854814
- Gene Name
- Gene Aliases
-
DRE3, YIL003W
- Chromosome Location
-
Chr.IX: 349122 - 350003 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179353.1 |
NP_012263.1 |
1 - 1 |
654 |
88 |
View Full Product Details