Assay Design
Gene Transcripts
- Gene Symbol
- YIA6
- Entrez Gene ID
-
854811
- Gene Name
- Gene Aliases
-
NDT1, YIL006W
- Chromosome Location
-
Chr.IX: 344062 - 345183 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179356.1 |
NP_012260.1 |
1 - 1 |
1052 |
109 |
View Full Product Details