Assay Design
Gene Transcripts
- Gene Symbol
- MND2
- Entrez Gene ID
-
854843
- Gene Name
- Gene Aliases
-
YIR025W
- Chromosome Location
-
Chr.IX: 403659 - 404765 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179547.1 |
NP_012291.1 |
1 - 1 |
1046 |
87 |
View Full Product Details