Assay Design
Gene Transcripts
- Gene Symbol
- DCG1
- Entrez Gene ID
-
854848
- Gene Name
- Gene Aliases
-
YIR030C
- Chromosome Location
-
Chr.IX: 412036 - 412770 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179552.1 |
NP_012296.1 |
1 - 1 |
650 |
81 |
View Full Product Details