Assay Design
Gene Transcripts
- Gene Symbol
- TRK1
- Entrez Gene ID
-
853312
- Gene Name
- Gene Aliases
-
YJL129C
- Chromosome Location
-
Chr.X: 173599 - 177306 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181562.1 |
NP_012406.1 |
1 - 1 |
3655 |
89 |
View Full Product Details