Assay Design
Gene Transcripts
- Gene Symbol
- JJJ2
- Entrez Gene ID
-
853277
- Gene Name
- Gene Aliases
-
YJL162C
- Chromosome Location
-
Chr.X: 113878 - 115629 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181595.1 |
NP_012373.2 |
1 - 1 |
1447 |
99 |
View Full Product Details