Assay Design
Gene Transcripts
- Gene Symbol
- SWE1
- Entrez Gene ID
-
853252
- Gene Name
- Gene Aliases
-
WEE1, YJL187C
- Chromosome Location
-
Chr.X: 76803 - 79262 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181620.1 |
NP_012348.1 |
1 - 1 |
2353 |
98 |
View Full Product Details