Assay Design
Gene Transcripts
- Gene Symbol
- ELO1
- Entrez Gene ID
-
853243
- Gene Name
- Gene Aliases
-
YJL196C
- Chromosome Location
-
Chr.X: 67850 - 68782 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181629.1 |
NP_012339.1 |
1 - 1 |
755 |
93 |
View Full Product Details