Assay Design
Gene Transcripts
- Gene Symbol
- CDC11
- Entrez Gene ID
-
853539
- Gene Name
- Gene Aliases
-
PSL9, YJR076C
- Chromosome Location
-
Chr.X: 575354 - 576601 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181734.1 |
NP_012610.1 |
1 - 1 |
1149 |
75 |
View Full Product Details