Assay Design
Gene Transcripts
- Gene Symbol
- AAD10
- Entrez Gene ID
-
853620
- Gene Name
- Gene Aliases
-
YJR155W
- Chromosome Location
-
Chr.X: 727405 - 728271 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181813.1 |
NP_012689.1 |
1 - 1 |
52 |
72 |
View Full Product Details