Assay Design
Gene Transcripts
- Gene Symbol
- UFD4
- Entrez Gene ID
-
853859
- Gene Name
- Gene Aliases
-
YKL010C
- Chromosome Location
-
Chr.XI: 421424 - 425875 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179576.3 |
NP_012915.3 |
1 - 1 |
4347 |
89 |
View Full Product Details