Assay Design
Gene Transcripts
- Gene Symbol
- SDH3
- Entrez Gene ID
-
853716
- Gene Name
- Gene Aliases
-
CYB3, YKL141W, YKL4
- Chromosome Location
-
Chr.XI: 179667 - 180263 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179707.1 |
NP_012781.1 |
1 - 1 |
448 |
67 |
View Full Product Details